Merck
  • Metformin ameliorates sepsis-induced brain injury by inhibiting apoptosis, oxidative stress and neuroinflammation via the PI3K/Akt signaling pathway.
Metformin ameliorates sepsis-induced brain injury by inhibiting apoptosis, oxidative stress and neuroinflammation via the PI3K/Akt signaling pathway.
Oncotarget (2017-12-13)
Guangming Tang, Huiyun Yang, Jing Chen, Mengrao Shi, Lingqing Ge, Xuhua Ge, Guoji Zhu
ABSTRACT
Sepsis-induced brain injuries increase mortality, morbidity, cognitive impairment and lack of effective therapeutic treatment. Previous studies have suggested that metformin provides neuroprotective effects against ischemia, brain trauma and other brain damage, but whether metformin protects a septic brain remains unknown. Thus, the aim of this study is to investigate the possible effects and the mechanism of metformin against septic brain damage using the cecal ligation and puncture (CLP) model. Mice were randomly divided into five groups: the Sham group, CLP group, CLP+ Met group, CLP+ vehicle group and CLP+ Met+ LY group. The survival percentage and brain water content were examined, and the Morris water maze was conducted to determine the protective effect of metformin. Neuronal apoptosis in the cerebral cortex, striatum and hippocampus was examined using TUNEL assay and immunohistochemistry, and western blot was applied to measure the expression of p-Akt. The results indicate that metformin can increase survival percentage, decrease brain edema, preserve the blood-brain barrier (BBB) and improve cognitive function. Metformin also reduced the neuronal apoptosis induced by sepsis and increased the phosphorylation of Akt. However, the protective effect of metformin can be reversed by LY294002, a PI3K inhibitor. In summary, our results demonstrate that metformin can exert a neuroprotective effect by activating the PI3K/Akt signaling pathway.
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF488A F(ab′)2 fragment of antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution