Merck
  • Effect of Annexin A1 gene on the proliferation and invasion of esophageal squamous cell carcinoma cells and its regulatory mechanisms.
Effect of Annexin A1 gene on the proliferation and invasion of esophageal squamous cell carcinoma cells and its regulatory mechanisms.
International journal of molecular medicine (2016-12-31)
Gaohua Han, Kaijin Lu, Junxing Huang, Jun Ye, Shengbin Dai, Yunyao Ye, Lixin Zhang
ABSTRACT
The aim of this study was to examine the effect of Annexin A1 (ANXA1) on the proliferation, migration and invasion of esophageal squamous cell carcinoma (ESCC) cells and its possible mechanisms of action. After constructing the ANXA1 overexpression plasmid, we transfected this plasmid and/or microRNA (miRNA)‑196a mimic into ESCC cells (Eca109 cell line). Methyl thiazolyl tetrazolium (MTT) assay and Transwell chamber assay were performed to determine cell proliferation, migration and invasion, respectively. Western blot analysis was used to examine the protein expression levels of ANXA1, Snail and E-cadherin. RT-PCR was used to detect the expression of miRNA-196a. Our results revealed that ANXA1 expression was upregulated in the cells transfected with the ANXA1 overexpression plasmid, and cell proliferation, migration and invasion were significantly increased (p=0.004, p<0.001 and p=0.011, respectively). In the cells transfected with the miRNA‑196a mimic, miRNA‑196a expression was significantly upregulated (p<0.001). However, miRNA-196a expression was downregulated in the cells transfected with the ANXA1 overexpression plasmid. In addition, in the cells transfected with the miRNA‑196a mimic, cell proliferation, migration and invasion were significantly decreased (p=0.027, p=0.009 and p=0.021, respectively). In the cells transfected with the ANXA1 overexpression plasmid, the expression of Snail was upregulated and that of E-cadherin was downregulated. However, the opposite was observed in the cells transfected with the miRNA‑196a mimic. Our findings thus demonstrate that ANXA1 promotes the proliferation of Eca109 cells, and increases the expression of Snail, whereas it inhibits that of E-cadherin, thus enhancing the migration and invasion of ESCC cells. miRNA-196a negatively regulates the expression of ANXA1, thereby inhibiting the proliferation, invasion and metastasis of ESCC cells.
Sigma-Aldrich
Monoclonal Anti-SNAI2 antibody produced in mouse, clone 3C12, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.