Merck
  • Computational Design and Synthesis of Novel Fluoro-Analogs of Combretastatins A-4 and A-1.
Computational Design and Synthesis of Novel Fluoro-Analogs of Combretastatins A-4 and A-1.
Journal of fluorine chemistry (2018-01-10)
Yao Zong, Christie Shea, Katherine Maffucci, Iwao Ojima
ABSTRACT
Combretastatin A-1 (CA-1) and combretastatin A-4 (CA-4) isolated from the African bush willow
Sigma-Aldrich
4-(Difluoromethoxy)benzaldehyde, 95%
Pricing and availability is not currently available.