Merck
Microbes inside--from diversity to function: the case of Akkermansia.
The ISME journal (2012-03-23)
Clara Belzer, Willem M de Vos
ABSTRACT
The human intestinal tract is colonized by a myriad of microbes that have developed intimate interactions with the host. In healthy individuals, this complex ecosystem remains stable and resilient to stressors. There is significant attention on the understanding of the composition and function of this intestinal microbiota in health and disease. Current developments in metaomics and systems biology approaches allow to probe the functional potential and activity of the intestinal microbiota. However, all these approaches inherently suffer from the fact that the information on macromolecules (DNA, RNA and protein) is collected at the ecosystem level. Similarly, all physiological and other information collected from isolated strains relates to pure cultures grown in vitro or in gnotobiotic systems. It is essential to integrate these two worlds of predominantly chemistry and biology by linking the molecules to the cells. Here, we will address the integration of omics- and culture-based approaches with the complexity of the human intestinal microbiota in mind and the mucus-degrading bacteria Akkermansia spp. as a paradigm.
Sigma-Aldrich
Microbial DNA standard from Akkermansia muciniphila, Suitable for PCR, sequencing and NGS, 10 ng/μL
Sigma-Aldrich
Akkermansia muciniphila FISH probe-ATTO488, Probe for fluorescence in situ hybridization (FISH), 20µM in water