Merck
  • Coupling sweeping-micellar electrokinetic chromatography with tandem mass spectrometry for the therapeutic monitoring of benzimidazoles in animal urine by dilute and shoot.
Coupling sweeping-micellar electrokinetic chromatography with tandem mass spectrometry for the therapeutic monitoring of benzimidazoles in animal urine by dilute and shoot.
Talanta (2017-08-27)
Carmen Tejada-Casado, David Moreno-González, Monsalud Del Olmo-Iruela, Ana M García-Campaña, Francisco J Lara
ABSTRACT
A new method based on micellar electrokinetic chromatography-tandem mass spectrometry (MEKC-MS/MS) has been developed for the identification and simultaneous quantification of thirteen benzimidazoles in animal urine. In order to obtain an appropriate separation with the highest sensitivity, different electrophoretic parameters were evaluated. Under optimum conditions, the separation was performed using ammonium perfluorooctanoate as volatile surfactant and electrophoretic buffer (50mM, pH 9). To increase the sensitivity, a stacking mode named sweeping was applied, using water as injection solvent at 50mbar for 75s, obtaining sensitivity enhancement factors from 50 to 181. The method was applied to different animal urine samples, including sheep, cow and goat. The sample treatment consisted of a 1:10 (v/v) dilution with water. Calibration using sheep urine samples can be used for both goat and cow urine samples with a relative bias below 25% and relative standard deviations lower than 8%. The limits of detection were below 70µgL
Product Description
Supelco
Fenbendazole sulfone, VETRANAL®, analytical standard
Supelco
Mebendazole-amine, VETRANAL®, analytical standard
Supelco
5-Hydroxymebendazole, VETRANAL®, analytical standard
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Supelco
Albendazole sulfone, VETRANAL®, analytical standard