Merck
  • Functionalized metal organic frameworks for effective capture of radioactive organic iodides.
Functionalized metal organic frameworks for effective capture of radioactive organic iodides.
Faraday discussions (2017-06-28)
Baiyan Li, Xinglong Dong, Hao Wang, Dingxuan Ma, Kui Tan, Zhan Shi, Yves J Chabal, Yu Han, Jing Li
ABSTRACT
Highly efficient capture of radioactive organic iodides (ROIs) from off-gas mixtures remains a substantial challenge for nuclear waste treatment. Current materials utilized for ROI sequestration suffer from low capacity, high cost (e.g. use of noble metals), and poor recyclability. Recently, we have developed a new strategy to tackle this challenge by functionalizing MOF materials with tertiary amines to create molecular traps for the effective capture and removal of ROIs (e.g. radioactive methyl iodide) from nuclear wastes. To further enhance the uptake capacity and performance of CH
Sigma-Aldrich
3-(Dimethylamino)-1-propylamine, 99%