Merck
  • Downregulation of X-linked inhibitor of apoptosis protein by '7-Benzylidenenaltrexone maleate' sensitizes pancreatic cancer cells to TRAIL-induced apoptosis.
Downregulation of X-linked inhibitor of apoptosis protein by '7-Benzylidenenaltrexone maleate' sensitizes pancreatic cancer cells to TRAIL-induced apoptosis.
Oncotarget (2017-10-06)
So Young Kim, Sojung Park, SeonA Yoo, Jin Kyung Rho, Eun Sung Jun, Suhwan Chang, Kyung Kon Kim, Song Cheol Kim, Inki Kim
ABSTRACT
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a potential biological anticancer agent. However, a wide range of human primary cancers, including pancreatic cancer, display resistance to apoptosis induction by TRAIL. Therefore, this resistance needs to be overcome to allow TRAIL to be successfully used in cancer therapy. In this study, we performed a compound screen to isolate TRAIL sensitizers and found that one of the identified compounds, 7-benzylidenenaltrexone maleate (BNTX), sensitized pancreatic cancer cells to TRAIL-induced apoptotic cell death. The combination of BNTX with TRAIL promoted the release of cytochrome
Sigma-Aldrich
MISSION® esiRNA, targeting human XIAP
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-phospho-XIAP (pSer87) antibody produced in rabbit, affinity isolated antibody