Merck
  • Identification and Characterization of Daurichromenic Acid Synthase Active in Anti-HIV Biosynthesis.
Identification and Characterization of Daurichromenic Acid Synthase Active in Anti-HIV Biosynthesis.
Plant physiology (2017-07-07)
Miu Iijima, Ryosuke Munakata, Hironobu Takahashi, Hiromichi Kenmoku, Ryuichi Nakagawa, Takeshi Kodama, Yoshinori Asakawa, Ikuro Abe, Kazufumi Yazaki, Fumiya Kurosaki, Futoshi Taura
ABSTRACT
Daurichromenic acid (DCA) synthase catalyzes the oxidative cyclization of grifolic acid to produce DCA, an anti-HIV meroterpenoid isolated from
Sigma-Aldrich
Geranylgeraniol, ≥85% (GC)