Merck
Synthesis, antiviral evaluation and molecular docking studies of N
Bioorganic & medicinal chemistry (2017-06-11)
María C Soraires Santacruz, Matías Fabiani, Eliana F Castro, Lucía V Cavallaro, Liliana M Finkielsztein
ABSTRACT
A series of N
Sigma-Aldrich
5,6-Dimethoxy-1-indanone, 97%
Sigma-Aldrich
4-Methoxyphenyl isothiocyanate, 98%
Sigma-Aldrich
4-Nitrophenyl isothiocyanate, 98%
Sigma-Aldrich
4-Fluorophenyl isothiocyanate, 98%