- Synthesis and investigation of anticancer potential of radiolabeled naphthalene monoimide bearing imidazolium salt.
Synthesis and investigation of anticancer potential of radiolabeled naphthalene monoimide bearing imidazolium salt.
Chemical biology & drug design (2017-01-10)
Fatma Yurt Lambrecht, Kasim Ocakoglu, Suleyman Gokhan Colak, Onur Alp Ersoz, Ozge Er
PMID28067001
ABSTRACT
Imidazolium salts and derivatives have antitumor efficacy and toxic effects in different micro-organisms. In this study, an imidazolium bromide salt (NMI) was synthesized, and its antitumor potential was investigated by in vitro studies. Radiolabeling of synthesized NMI was carried out by iodogen method using