Merck
  • The CUL3-SPOP-DAXX axis is a novel regulator of VEGFR2 expression in vascular endothelial cells.
The CUL3-SPOP-DAXX axis is a novel regulator of VEGFR2 expression in vascular endothelial cells.
Scientific reports (2017-02-22)
Tomohisa Sakaue, Iori Sakakibara, Takahiro Uesugi, Ayako Fujisaki, Koh-Ichi Nakashiro, Hiroyuki Hamakawa, Eiji Kubota, Takashi Joh, Yu-Ki Imai, Hironori Izutani, Shigeki Higashiyama
ABSTRACT
Vascular endothelial cell growth factor receptor 2 (VEGFR2) is an essential receptor for the homeostasis of endothelial cells. In this study, we showed that NEDD8-conjugated Cullin3 (CUL3)-based ubiquitin E3 (UbE3) ligase plays a crucial role in VEGFR2 mRNA expression. Human umbilical vein endothelial cells treated with MLN4924, an inhibitor of NEDD8-activating enzyme, or with CUL3 siRNA drastically lost their response to VEGF due to the intense decrease in VEGFR2 expression. Moreover, speckle-type POZ protein (SPOP) and death-domain associated protein (DAXX) were involved in the CUL3 UbE3 ligase complex as a substrate adaptor and a substrate, respectively. Knockdown of SPOP and CUL3 led to the upregulation of DAXX protein and downregulation of VEGFR2 levels. These levels were inversely correlated with one another. In addition, simultaneous knockdown of SPOP and DAXX completely reversed the downregulation of VEGFR2 levels. Moreover, the CUL3-SPOP-DAXX axis had the same effects on NOTCH1, DLL4 and NRP1 expression. Taken together, these findings suggest that the CUL3-SPOP-DAXX axis plays a very important role in endothelial cell function by targeting key angiogenic regulators.
Sigma-Aldrich
MISSION® esiRNA, targeting human CUL3
Sigma-Aldrich
Anti-Cullin 3 antibody ,Mouse monoclonal, clone CUL3-9, purified from hybridoma cell culture