Merck
  • Dedicated SNAREs and specialized TRIM cargo receptors mediate secretory autophagy.
Dedicated SNAREs and specialized TRIM cargo receptors mediate secretory autophagy.
The EMBO journal (2016-12-10)
Tomonori Kimura, Jingyue Jia, Suresh Kumar, Seong Won Choi, Yuexi Gu, Michal Mudd, Nicolas Dupont, Shanya Jiang, Ryan Peters, Farzin Farzam, Ashish Jain, Keith A Lidke, Christopher M Adams, Terje Johansen, Vojo Deretic
ABSTRACT
Autophagy is a process delivering cytoplasmic components to lysosomes for degradation. Autophagy may, however, play a role in unconventional secretion of leaderless cytosolic proteins. How secretory autophagy diverges from degradative autophagy remains unclear. Here we show that in response to lysosomal damage, the prototypical cytosolic secretory autophagy cargo IL-1β is recognized by specialized secretory autophagy cargo receptor TRIM16 and that this receptor interacts with the R-SNARE Sec22b to recruit cargo to the LC3-II+ sequestration membranes. Cargo secretion is unaffected by downregulation of syntaxin 17, a SNARE promoting autophagosome-lysosome fusion and cargo degradation. Instead, Sec22b in combination with plasma membrane syntaxin 3 and syntaxin 4 as well as SNAP-23 and SNAP-29 completes cargo secretion. Thus, secretory autophagy utilizes a specialized cytosolic cargo receptor and a dedicated SNARE system. Other unconventionally secreted cargo, such as ferritin, is secreted via the same pathway.
Sigma-Aldrich
Pararosaniline Solution
Sigma-Aldrich
Human Ferritin ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Monoclonal Anti-NCOA4, (C-terminal) antibody produced in mouse, clone 1F11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-STX17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution