Merck
  • The tumor microenvironment underlies acquired resistance to CSF-1R inhibition in gliomas.
The tumor microenvironment underlies acquired resistance to CSF-1R inhibition in gliomas.
Science (New York, N.Y.) (2016-05-21)
Daniela F Quail, Robert L Bowman, Leila Akkari, Marsha L Quick, Alberto J Schuhmacher, Jason T Huse, Eric C Holland, James C Sutton, Johanna A Joyce
ABSTRACT
Macrophages accumulate with glioblastoma multiforme (GBM) progression and can be targeted via inhibition of colony-stimulating factor-1 receptor (CSF-1R) to regress high-grade tumors in animal models of this cancer. However, whether and how resistance emerges in response to sustained CSF-1R blockade is unknown. We show that although overall survival is significantly prolonged, tumors recur in >50% of mice. Gliomas reestablish sensitivity to CSF-1R inhibition upon transplantation, indicating that resistance is tumor microenvironment-driven. Phosphatidylinositol 3-kinase (PI3K) pathway activity was elevated in recurrent GBM, driven by macrophage-derived insulin-like growth factor-1 (IGF-1) and tumor cell IGF-1 receptor (IGF-1R). Combining IGF-1R or PI3K blockade with CSF-1R inhibition in recurrent tumors significantly prolonged overall survival. Our findings thus reveal a potential therapeutic approach for treating resistance to CSF-1R inhibitors.
Sigma-Aldrich
AS1517499, ≥98% (HPLC)
Pricing and availability is not currently available.