Merck
  • Evaluation of CITED1, SIX1, and CD56 protein expression for identification of blastemal elements in Wilms tumor.
Evaluation of CITED1, SIX1, and CD56 protein expression for identification of blastemal elements in Wilms tumor.
American journal of clinical pathology (2014-05-20)
Daniel Sehic, Cristina D Ciornei, David Gisselsson
ABSTRACT
Successful further treatment of Wilms tumors (WTs) after preoperative chemotherapy and surgery depends on correct histopathologic risk stratification, including quantification of remaining blastemal elements. In the present study, we assessed the usefulness of protein markers for the detection of WT blastema. Expression of the candidate blastemal protein markers CITED1, SIX1, and CD56 was evaluated by immunofluorescence regarding sensitivity and specificity for staining blastema in a tissue microarray containing cores from 30 WTs, a small number of rarer pediatric renal neoplasms, and normal postnatal kidney. CITED1, SIX1, and CD56 were expressed in blastema in 100%, 89%, and 74%, respectively, of the WTs with this component present. However, they were also expressed in 64%, 25%, and 79%, respectively, of epithelial WT elements and 48%, 52%, and 62%, respectively, of stromal WT elements. SIX1 showed the highest specificity, CITED1 the highest sensitivity, and CD56 low specificity and sensitivity for detection of postchemotherapy WT blastema. Cytokeratin staining proved to be a useful way to determine rudimentary tubular elements not readily recognized by routine staining.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SIX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Cytokeratin AE1/AE3 Antibody, recognizes acidic & basic cytokeratins, clone AE1/AE3, clone AE1/AE3, Chemicon®, from mouse