Merck
  • Reducing GBA2 Activity Ameliorates Neuropathology in Niemann-Pick Type C Mice.
Reducing GBA2 Activity Ameliorates Neuropathology in Niemann-Pick Type C Mice.
PloS one (2015-08-15)
André R A Marques, Jan Aten, Roelof Ottenhoff, Cindy P A A van Roomen, Daniela Herrera Moro, Nike Claessen, María Fernanda Vinueza Veloz, Kuikui Zhou, Zhanmin Lin, Mina Mirzaian, Rolf G Boot, Chris I De Zeeuw, Herman S Overkleeft, Yildiz Yildiz, Johannes M F G Aerts
ABSTRACT
The enzyme glucocerebrosidase (GBA) hydrolyses glucosylceramide (GlcCer) in lysosomes. Markedly reduced GBA activity is associated with severe manifestations of Gaucher disease including neurological involvement. Mutations in the GBA gene have recently also been identified as major genetic risk factor for Parkinsonism. Disturbed metabolism of GlcCer may therefore play a role in neuropathology. Besides lysosomal GBA, cells also contain a non-lysosomal glucosylceramidase (GBA2). Given that the two β-glucosidases share substrates, we speculated that over-activity of GBA2 during severe GBA impairment might influence neuropathology. This hypothesis was studied in Niemann-Pick type C (Npc1-/-) mice showing secondary deficiency in GBA in various tissues. Here we report that GBA2 activity is indeed increased in the brain of Npc1-/- mice. We found that GBA2 is particularly abundant in Purkinje cells (PCs), one of the most affected neuronal populations in NPC disease. Inhibiting GBA2 in Npc1-/- mice with a brain-permeable low nanomolar inhibitor significantly improved motor coordination and extended lifespan in the absence of correction in cholesterol and ganglioside abnormalities. This trend was recapitulated, although not to full extent, by introducing a genetic loss of GBA2 in Npc1-/- mice. Our findings point to GBA2 activity as therapeutic target in NPC.
Sigma-Aldrich
Anti-Glucocerebrosidase (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Calbindin D-28K (Ab-1) Rabbit pAb, lyophilized, Calbiochem®