Merck
  • Stimulation of autophagy by rapamycin protects neurons from remote degeneration after acute focal brain damage.
Stimulation of autophagy by rapamycin protects neurons from remote degeneration after acute focal brain damage.
Autophagy (2012-01-18)
Maria Teresa Viscomi, Marcello D'Amelio, Virve Cavallucci, Laura Latini, Elisa Bisicchia, Francesca Nazio, Francesca Fanelli, Mauro Maccarrone, Sandra Moreno, Francesco Cecconi, Marco Molinari
ABSTRACT
Autophagy is the evolutionarily conserved degradation and recycling of cellular constituents. In mammals, autophagy is implicated in the pathogenesis of many neurodegenerative diseases. However, its involvement in acute brain damage is unknown. This study addresses the function of autophagy in neurodegeneration that has been induced by acute focal cerebellar lesions. We provide morphological, ultrastructural, and biochemical evidence that lesions in a cerebellar hemisphere activate autophagy in axotomized precerebellar neurons. Through time course analyses of the apoptotic cascade, we determined mitochondrial dysfunction to be the early trigger of degeneration. Further, the stimulation of autophagy by rapamycin and the employment of mice with impaired autophagic responses allowed us to demonstrate that autophagy protects from damage promoting functional recovery. These findings have therapeutic significance, demonstrating the potential of pro-autophagy treatments for acute brain pathologies, such as stroke and brain trauma.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.
Sigma-Aldrich
AptaFluor® SAH Methyltransferase Assay