Merck
  • A role for the mitochondrial-associated protein p32 in regulation of trophoblast proliferation.
A role for the mitochondrial-associated protein p32 in regulation of trophoblast proliferation.
Molecular human reproduction (2014-05-31)
P Matos, J A Horn, F Beards, S Lui, M Desforges, L K Harris
ABSTRACT
p32 is a conserved eukaryotic protein which is primarily expressed in the mitochondria and regulates cell proliferation, migration and metabolism in various tissues. In this study, we sought to examine the expression and function of p32 in the human placenta. p32 was highly expressed in the syncytiotrophoblast, the underlying cytotrophoblast (CTB), the vascular endothelium and by a proportion of cells in the villous stroma in first trimester and term placenta. p32 mRNA and protein expression was significantly higher in the first trimester of pregnancy than at term, and expression in the trophoblast was significantly reduced in placentas from women with fetal growth restriction (FGR). Small interfering RNA (siRNA)-mediated knockdown of p32 in term placental explants significantly reduced the number of Ki67-positive CTB, but did not alter CTB apoptosis or necrosis. p32 knockdown increased lactate production, reduced glucose extraction from culture medium and was associated with reduced MitoTracker dye accumulation in trophoblast mitochondria. p32 knockdown was also associated with a significant reduction in expression of the mitochondrial respiratory complexes I and IV. These data suggest that p32 expression is important for CTB proliferation, via a mechanism involving regulation of normal mitochondrial function. As p32 expression is reduced in FGR placentas, this may contribute to some of the observed placental pathology, such as reduced CTB proliferation and mitochondrial dysfunction.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from mouse serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
Anti-C1QBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution