Merck
  • Identification of nucleolin as a lipid-raft-dependent β1-integrin-interacting protein in A375 cell migration.
Identification of nucleolin as a lipid-raft-dependent β1-integrin-interacting protein in A375 cell migration.
Molecules and cells (2013-12-03)
Jiajia Bi, Ruifei Wang, Yue Zhang, Xiaoqing Han, Khamal Kwesi Ampah, Wenguang Liu, Xianlu Zeng
ABSTRACT
Lipid rafts are related to cell surface receptor function. Integrin is a major surface receptor protein in cell adhesion and migration on the extracellular matrix (ECM). Here, we showed that lipid rafts played a critical role in human melanoma A375 cell spreading and migration on fibronectin; an important component of the ECM that interacts with β1 integrin. We found that the disruption of lipid rafts did not markedly inhibit the expression and activation of β1 integrin. By coimmunoprecipitation and mass spectrometry, we investigated the influence of lipid rafts on the β1 integrin complex and identified nucleolin as a potential lipid-raft-dependent β1-integrin-interacting protein. Upon confirmation of the interaction between β1 integrin and nucleolin, further studies revealed that nucleolin colocalized with β1 integrin in lipid rafts and raft disruption interrupted their association. In addition, knockdown of nucleolin markedly attenuated A375 cell spreading and migration on fibronectin. Taken together, we demonstrated that nucleolin is a critical lipid-raft-dependent β1-integrin-interacting protein in A375 cell spreading and migration on fibronectin.
Product Description
Millipore
Anti-Integrin β1 Antibody, clone TDM29, clone TDM29, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free, clone HUTS-4, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid