Merck
A natural light-driven inward proton pump.
Nature communications (2016-11-18)
Keiichi Inoue, Shota Ito, Yoshitaka Kato, Yurika Nomura, Mikihiro Shibata, Takayuki Uchihashi, Satoshi P Tsunoda, Hideki Kandori
ABSTRACT
Light-driven outward H
Sigma-Aldrich
Membrane Scaffold Protein 1E3D1, recombinant, expressed in E. coli
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Soy Phospholipids, mixture of phospholipids
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
all trans-Retinal, powder, ≥98%