Merck
  • Estradiol Reduces Connexin43 Gap Junctions in the Uterus during Adenomyosis in Cows.
Estradiol Reduces Connexin43 Gap Junctions in the Uterus during Adenomyosis in Cows.
Polish journal of veterinary sciences (2016-10-21)
A J Korzekwa, M Łupicka, B M Socha, A A Szczepańska
ABSTRACT
Adenomyosis is defined as the presence of glandular foci external to the endometrium of the uterus, either in the myometrium or/and perimetrium, depending on the progress of this dysfunction. To date, we showed that steroids secretion and prolactin expression and proliferative processes are disturbed during uterine adenomyosis in cows. During endometriosis in eutopic endometrium in women, gap junctions are down regulated. The transmembrane gap junction protein, connexin (Cx43) is necessary for endometrial morphological, biochemical and angiogenic functions. The aim of this study is recognition of adenomyosis etiology by determination of the role of Cx43 in this process. Immunolocalization and comparison of Cx43 mRNA and protein expression in healthy (N=9) and adenomyotic uterine tissue (N=9), and Cx43 mRNA expression (real time PCR) in uterine stromal - myometrium co-culture under 24-hour stimulation with 17-beta estradiol (10-7M) isolated from healthy (N=5) and adenomyotic (N=5) cows were determined. Cx43 was localized in healthy and adenomyotic uteri. mRNA and protein expression was down-regulated in uterine tissue in adenomyotic compared with healthy cows (p<0.05). Estradiol stimulated Cx43 mRNA expression in myometrial cell culture and co-culture of stromal and myometrial cells in adenomyotic compared with healthy cows (p<0.05). In summary, down-regulation of Cx43 expression in the junction zone might play an important role in pathogenesis of adenomyosis. Estradiol modulates gap junctions during adenomyosis.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Medium 199, HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture