Merck
  • Neural ablation of the PARK10 candidate Plpp3 leads to dopaminergic transmission deficits without neurodegeneration.
Neural ablation of the PARK10 candidate Plpp3 leads to dopaminergic transmission deficits without neurodegeneration.
Scientific reports (2016-04-12)
Sandra Gómez-López, Ana Valeria Martínez-Silva, Teresa Montiel, Daniel Osorio-Gómez, Federico Bermúdez-Rattoni, Lourdes Massieu, Diana Escalante-Alcalde
ABSTRACT
Parkinson's disease (PD) is a multifactorial neurodegenerative disorder, characterised by the progressive loss of midbrain dopaminergic neurons and a variety of motor symptoms. The gene coding for the phospholipid phosphatase 3, PLPP3 (formerly PPAP2B or LPP3), maps within the PARK10 locus, a region that has been linked with increased risk to late-onset PD. PLPP3 modulates the levels of a range of bioactive lipids controlling fundamental cellular processes within the central nervous system. Here we show that PLPP3 is enriched in astroglial cells of the adult murine ventral midbrain. Conditional inactivation of Plpp3 using a Nestin::Cre driver results in reduced mesencephalic levels of sphingosine-1-phosphate receptor 1 (S1P1), a well-known mediator of pro-survival responses. Yet, adult PLPP3-deficient mice exhibited no alterations in the number of dopaminergic neurons or in the basal levels of striatal extracellular dopamine (DA). Potassium-evoked DA overflow in the striatum, however, was significantly decreased in mutant mice. Locomotor evaluation revealed that, although PLPP3-deficient mice exhibit motor impairment, this is not progressive or responsive to acute L-DOPA therapy. These findings suggest that disruption of Plpp3 during early neural development leads to dopaminergic transmission deficits in the absence of nigrostriatal degeneration, and without causing an age-related locomotor decline consistent with PD.
Sigma-Aldrich
Anti-GLAST Antibody, clone 8C11.1, clone 8C11.1, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit