Merck
  • Catalytic asymmetric aminohydroxylation provides a short taxol side-chain synthesis.
Catalytic asymmetric aminohydroxylation provides a short taxol side-chain synthesis.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989) (1996-08-01)
G Li, K B Sharpless
ABSTRACT
The p-toluenesulfonamide derivate of the C-13 side-chain of taxol was prepared on a one third mole scale in a single step from methyl cinnamate. The process employed is catalytic asymmetric aminohydroxylation (catalytic AA). In the present case, there is no work-up other than filtration of the pure product which is insoluble in the reaction mixture. The sulfonamide protecting group is removed by acidic hydrolysis.
Sigma-Aldrich
Chloramine T trihydrate, ACS reagent, 98%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl cinnamate, ≥99.0% (GC)
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
Chloramine-T hydrate, 95%
Sigma-Aldrich
Methyl trans-cinnamate, 99%
Sigma-Aldrich
Methyl cinnamate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methyl trans-cinnamate, ≥98%, stabilized, FCC, FG