Merck
  • Application of 6-nitrocoumarin as a substrate for the fluorescent detection of nitroreductase activity in Sporothrix schenckii.
Application of 6-nitrocoumarin as a substrate for the fluorescent detection of nitroreductase activity in Sporothrix schenckii.
Revista do Instituto de Medicina Tropical de Sao Paulo (2013-09-17)
Cheila Denise Ottonelli Stopiglia, Mariana Carissimi, Tatiane Caroline Daboit, Valter Stefani, Valeriano Antonio Corbellini, Maria Lúcia Scroferneker
ABSTRACT
Sporothrix schenckii is a thermal dimorphic pathogenic fungus causing a subcutaneous mycosis, sporotrichosis. Nitrocoumarin represents a fluorogenic substrate class where the microbial nitroreductase activity produces several derivatives, already used in several other enzyme assays. The objective of this study was the analysis of 6-nitrocoumarin (6-NC) as a substrate to study the nitroreductase activity in Sporothrix schenckii. Thirty-five samples of S. schenckii were cultivated for seven, 14 and 21 days at 35 °C in a microculture containing 6-nitrocoumarin or 6-aminocoumarin (6-AC) dissolved in dimethyl sulfoxide or dimethyl sulfoxide as a negative control, for posterior examination under an epifluorescence microscope. The organic layer of the seven, 14 and 21-day cultures was analyzed by means of direct illumination with 365 nm UV light and by means of elution on G silica gel plate with hexane:ethyl acetate 1:4 unveiled with UV light. All of the strains showed the presence of 6-AC (yellow fluorescence) and 6-hydroxylaminocoumarin (blue fluorescence) in thin layer chromatography, which explains the green fluorescence observed in the fungus structure. The nitroreductase activity is widely distributed in the S. schenckii complex and 6-NC is a fluorogenic substrate of easy access and applicability for the nitroreductase activity detection.