Merck
  • Therapeutic potential of in utero mesenchymal stem cell (MSCs) transplantation in rat foetuses with spina bifida aperta.
Therapeutic potential of in utero mesenchymal stem cell (MSCs) transplantation in rat foetuses with spina bifida aperta.
Journal of cellular and molecular medicine (2011-10-19)
Hui Li, Fei Gao, Lili Ma, Junhong Jiang, Jianing Miao, Mingyu Jiang, Yang Fan, Lili Wang, Di Wu, Bo Liu, Weilin Wang, Vincent Chi Hang Lui, Zhengwei Yuan
ABSTRACT
Neural tube defects (NTDs) are complex congenital malformations resulting from incomplete neurulation in embryo. Despite surgical repair of the defect, most of the patients who survive with NTDs have a multiple system handicap due to neuron deficiency of the defective spinal cord. In this study, we successfully devised a prenatal surgical approach and transplanted mesenchymal stem cells (MSCs) to foetal rat spinal column to treat retinoic acid induced NTDs in rat. Transplanted MSCs survived, grew and expressed markers of neurons, glia and myoblasts in the defective spinal cord. MSCs expressed and perhaps induced the surrounding spinal tissue to express neurotrophic factors. In addition, MSC reduced spinal tissue apoptosis in NTD. Our results suggested that prenatal MSC transplantation could treat spinal neuron deficiency in NTDs by the regeneration of neurons and reduced spinal neuron death in the defective spinal cord.
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse