Merck
  • The fungal fast lane: common mycorrhizal networks extend bioactive zones of allelochemicals in soils.
The fungal fast lane: common mycorrhizal networks extend bioactive zones of allelochemicals in soils.
PloS one (2011-11-24)
E Kathryn Barto, Monika Hilker, Frank Müller, Brian K Mohney, Jeffrey D Weidenhamer, Matthias C Rillig
ABSTRACT
Allelopathy, a phenomenon where compounds produced by one plant limit the growth of surrounding plants, is a controversially discussed factor in plant-plant interactions with great significance for plant community structure. Common mycorrhizal networks (CMNs) form belowground networks that interconnect multiple plant species; yet these networks are typically ignored in studies of allelopathy. We tested the hypothesis that CMNs facilitate transport of allelochemicals from supplier to target plants, thereby affecting allelopathic interactions. We analyzed accumulation of a model allelopathic substance, the herbicide imazamox, and two allelopathic thiophenes released from Tagetes tenuifolia roots, by diffusion through soil and CMNs. We also conducted bioassays to determine how the accumulated substances affected plant growth. All compounds accumulated to greater levels in target soils with CMNs as opposed to soils without CMNs. This increased accumulation was associated with reduced growth of target plants in soils with CMNs. Our results show that CMNs support transfer of allelochemicals from supplier to target plants and thus lead to allelochemical accumulation at levels that could not be reached by diffusion through soil alone. We conclude that CMNs expand the bioactive zones of allelochemicals in natural environments, with significant implications for interspecies chemical interactions in plant communities.
Product Description
Supelco
Imazamox, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
1 Estimated to ship on March 08, 2025
$136.00
5 mL
1 Estimated to ship on March 08, 2025
$460.00