Merck
  • Enhancing production and cytotoxic activity of polymeric soluble FasL-based chimeric proteins by concomitant expression of soluble FasL.
Enhancing production and cytotoxic activity of polymeric soluble FasL-based chimeric proteins by concomitant expression of soluble FasL.
PloS one (2013-08-31)
Aurore Morello, Sophie Daburon, Michel Castroviejo, Jean-François Moreau, Julie Dechanet-Merville, Jean-Luc Taupin
ABSTRACT
Membrane FasL is the natural trigger of Fas-mediated apoptosis. A soluble homotrimeric counterpart (sFasL) also exists which is very weakly active, and needs oligomerization beyond its trimeric state to induce apoptosis. We recently generated a soluble FasL chimera by fusing the immunoglobulin-like domain of the leukemia inhibitory factor receptor gp190 to the extracellular region of human FasL, which enabled spontaneous dodecameric homotypic polymerization of FasL. This polymeric soluble human FasL (pFasL) displayed anti-tumoral activity in vitro and in vivo without systemic cytotoxicity in mouse. In the present work, we focused on the improvement of pFasL, with two complementary objectives. First, we developed more complex pFasL-based chimeras that contained a cell-targeting module. Secondly, we attempted to improve the production and/or the specific activity of pFasL and of the cell-targeting chimeras. We designed two chimeras by fusing to pFasL the extracellular portions of the HLA-A2 molecule or of a human gamma-delta TCR, and analyzed the consequences of co-expressing these molecules or pFasL together with sFasL on their heterotopic cell production. This strategy significantly enhanced the production of pFasL and of the two chimeras, as well as the cytotoxic activity of the two chimeras but not of pFasL. These results provide the proof of concept for an optimization of FasL-based chimeric proteins for a therapeutic use.
Sigma-Aldrich
Monoclonal Anti-B2M-PE antibody produced in mouse, clone B2M-01, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)