Merck
  • MYH9 is an Essential Factor for Porcine Reproductive and Respiratory Syndrome Virus Infection.
MYH9 is an Essential Factor for Porcine Reproductive and Respiratory Syndrome Virus Infection.
Scientific reports (2016-04-27)
Jiming Gao, Shuqi Xiao, Yihong Xiao, Xiangpeng Wang, Chong Zhang, Qin Zhao, Yuchen Nan, Baicheng Huang, Hongliang Liu, Ningning Liu, Junhua Lv, Taofeng Du, Yani Sun, Yang Mu, Gang Wang, Shahid Faraz Syed, Gaiping Zhang, Julian A Hiscox, Ian Goodfellow, En-Min Zhou
ABSTRACT
Porcine reproductive and respiratory syndrome (PRRS) caused by the PRRS virus (PRRSV) is an important swine disease worldwide. PRRSV has a limited tropism for certain cells, which may at least in part be attributed to the expression of the necessary cellular molecules serving as the virus receptors or factors on host cells for virus binding or entry. However, these molecules conferring PRRSV infection have not been fully characterized. Here we show the identification of non-muscle myosin heavy chain 9 (MYH9) as an essential factor for PRRSV infection using the anti-idiotypic antibody specific to the PRRSV glycoprotein GP5. MYH9 physically interacts with the PRRSV GP5 protein via its C-terminal domain and confers susceptibility of cells to PRRSV infection. These findings indicate that MYH9 is an essential factor for PRRSV infection and provide new insights into PRRSV-host interactions and viral entry, potentially facilitating development of control strategies for this important swine disease.
Sigma-Aldrich
Anti-Myosin IIB, non-muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-6X His IgG, CF770 antibody produced in mouse, ~2 mg/mL, clone 33D10.D2.G8, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)