Merck
  • Palladium-catalyzed, ligand-free Suzuki reaction in water using aryl fluorosulfates.
Palladium-catalyzed, ligand-free Suzuki reaction in water using aryl fluorosulfates.
Organic letters (2015-04-10)
Qiaobin Liang, Ping Xing, Zuogang Huang, Jiajia Dong, K Barry Sharpless, Xiaoxian Li, Biao Jiang
ABSTRACT
Aryl fluorosulfates were prepared by a simple method and employed as coupling partners in the Suzuki-Miyaura reaction. The cross-coupling reactions were performed in water under air at room temperature without ligands or additives such as surfactants or phase-transfer reagents and proceeded smoothly to give excellent yields. Aryl fluorosulfates could also be used as alternatives to halides or triflates in other coupling reactions.
Sigma-Aldrich
2-cyanophenyl sulfofluoridate