Merck
  • Production of high-starch, low-glucose potatoes through over-expression of the metabolic regulator SnRK1.
Production of high-starch, low-glucose potatoes through over-expression of the metabolic regulator SnRK1.
Plant biotechnology journal (2006-12-21)
Rowan S McKibbin, Nira Muttucumaru, Matthew J Paul, Stephen J Powers, Michael M Burrell, Steve Coates, Patrick C Purcell, Axel Tiessen, Peter Geigenberger, Nigel G Halford
ABSTRACT
Transgenic potato (Solanum tuberosum cv. Prairie) lines were produced over-expressing a sucrose non-fermenting-1-related protein kinase-1 gene (SnRK1) under the control of a patatin (tuber-specific) promoter. SnRK1 activity in the tubers of three independent transgenic lines was increased by 55%-167% compared with that in the wild-type. Glucose levels were decreased, at 17%-56% of the levels of the wild-type, and the starch content showed an increase of 23%-30%. Sucrose and fructose levels in the tubers of the transgenic plants did not show a significant change. Northern analyses of genes encoding sucrose synthase and ADP-glucose pyrophosphorylase, two key enzymes involved in the biosynthetic pathway from sucrose to starch, showed that the expression of both was increased in tubers of the transgenic lines compared with the wild-type. In contrast, the expression of genes encoding two other enzymes of carbohydrate metabolism, alpha-amylase and sucrose phosphate synthase, showed no change. The activity of sucrose synthase and ADP-glucose pyrophosphorylase was also increased, by approximately 20%-60% and three- to five-fold, respectively, whereas the activity of hexokinase was unchanged. The results are consistent with a role for SnRK1 in regulating carbon flux through the storage pathway to starch biosynthesis. They emphasize the importance of SnRK1 in the regulation of carbohydrate metabolism and resource partitioning, and indicate a specific role for SnRK1 in the control of starch accumulation in potato tubers.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications