Merck
  • Two nested developmental waves demarcate a compartment boundary in the mouse lung.
Two nested developmental waves demarcate a compartment boundary in the mouse lung.
Nature communications (2014-06-01)
Denise Martinez Alanis, Daniel R Chang, Haruhiko Akiyama, Mark A Krasnow, Jichao Chen
ABSTRACT
The lung is a branched tubular network with two distinct compartments--the proximal conducting airways and the peripheral gas exchange region--separated by a discrete boundary termed the bronchoalveolar duct junction (BADJ). Here we image the developing mouse lung in three-dimensions (3D) and show that two nested developmental waves demarcate the BADJ under the control of a global hormonal signal. A first wave of branching morphogenesis progresses throughout embryonic development, generating branches for both compartments. A second wave of conducting airway differentiation follows the first wave but terminates earlier, specifying the proximal compartment and setting the BADJ. The second wave is terminated by a glucocorticoid signalling: premature activation or loss of glucocorticoid signalling causes a proximal or distal shift, respectively, in BADJ location. The results demonstrate a new mechanism of boundary formation in complex, 3D organs and provide new insights into glucocorticoid therapies for lung defects in premature birth.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Supelco
Methimazole, analytical standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Anti-Aquaporin 1 Antibody, 1.0 mg/mL, Chemicon®
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, ≥98% (HPLC)
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
6-Propyl-2-thiouracil, enzyme inhibitor
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)