Merck
  • The suppression of maternal-fetal leukemia inhibitory factor signal relay pathway by maternal immune activation impairs brain development in mice.
The suppression of maternal-fetal leukemia inhibitory factor signal relay pathway by maternal immune activation impairs brain development in mice.
PloS one (2015-06-05)
Tsuyoshi Tsukada, Eriko Simamura, Hiroki Shimada, Takuma Arai, Nobuaki Higashi, Takuya Akai, Hideaki Iizuka, Toshihisa Hatta
ABSTRACT
Recent studies in rodents suggest that maternal immune activation (MIA) by viral infection is associated with schizophrenia and autism in offspring. Although maternal IL-6 is though t to be a possible mediator relating MIA induced these neuropsychiatric disorders, the mechanism remains to be elucidated. Previously, we reported that the maternal leukemia inhibitory factor (LIF)-placental ACTH-fetal LIF signaling relay pathway (maternal-fetal LIF signal relay) promotes neurogenesis of fetal cerebrum in rats. Here we report that the maternal-fetal LIF signal relay in mice is suppressed by injection of polyriboinosinic-polyribocytidylic acid into dams, which induces MIA at 12.5 days post-coitum. Maternal IL-6 levels and gene expression of placental suppressor of cytokine signaling 3 (Socs3) increased according to the severity of MIA and gene expression of placental Socs3 correlated with maternal IL-6 levels. Furthermore, we show that MIA causes reduction of LIF level in the fetal cerebrospinal fluid, resulting in the decreased neurogenesis in the cerebrum. These findings suggest that maternal IL-6 interferes the maternal-fetal LIF signal relay by inducing SOCS3 in the placenta and leads to decreased neurogenesis.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Leukemia Inhibitory Factor human, LIF, recombinant, expressed in E. coli, 10 Ī¼g/ml, buffered aqueous solution (pH 7.4), suitable for cell culture
Sigma-Aldrich
Leukemia Inhibitory Factor human, animal component free, recombinant, expressed in rice, 500 μg/mL
Sigma-Aldrich
LIF human, Animal-component free, recombinant, expressed in E. coli, ā‰„98% (SDS-PAGE), ā‰„98% (HPLC)
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, LIF, recombinant, expressed in E. coli, 10 Ī¼g/ml, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Anti-Leukemia Inhibitory Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder