Merck
  • Inhibition of indoleamine 2,3-dioxygenase promotes vascular inflammation and increases atherosclerosis in Apoe-/- mice.
Inhibition of indoleamine 2,3-dioxygenase promotes vascular inflammation and increases atherosclerosis in Apoe-/- mice.
Cardiovascular research (2015-03-10)
Konstantinos A Polyzos, Olga Ovchinnikova, Martin Berg, Roland Baumgartner, Hanna Agardh, John Pirault, Anton Gisterå, Alice Assinger, Andres Laguna-Fernandez, Magnus Bäck, Göran K Hansson, Daniel F J Ketelhuth
ABSTRACT
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the first and rate-limiting enzyme of the kynurenine pathway of tryptophan (Trp) degradation, with immune regulation and anti-inflammatory mechanisms in different diseases. However, the role of IDO and the endogenous degradation of Trp have never been directly examined in atherosclerosis development. We used the IDO inhibitor 1-methyl-Trp (1-MT) to determine the role of IDO-mediated Trp metabolism in vascular inflammation and atherosclerosis. Apoe(-/-) mice were treated with 1-MT in drinking water for 8 weeks. Systemic IDO inhibition led to a significant increase in atherosclerotic lesions that were ∼58 and 54% larger in the aortic arch and root, respectively. 1-MT treatment enhanced vascular inflammation, up-regulated VCAM-1 and CCL2, and increased CD68 macrophage accumulation into the plaque. Notably, the rise in VCAM-1 expression was not limited to the plaque but also found in smooth muscle cells (SMCs) of the tunica media. Furthermore, we found that IDO-dependent Trp metabolism by SMCs regulates VCAM-1 expression, and that 1-MT-induced acceleration of atherosclerosis and vascular inflammation can be reversed by exogenous administration of the Trp metabolite 3-hydroxyanthranilic acid (3-HAA). IDO-mediated Trp metabolism regulates vascular inflammation and plaque formation in hypercholesterolaemic Apoe(-/-) mice. Our data establish that this pathway plays a major role in the pathological process of atherogenesis.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
1-Methyl-DL-tryptophan, 97%