Merck
  • Aging accelerates memory extinction and impairs memory restoration in Drosophila.
Aging accelerates memory extinction and impairs memory restoration in Drosophila.
Biochemical and biophysical research communications (2015-04-07)
Nannan Chen, Aike Guo, Yan Li
ABSTRACT
Age-related memory impairment (AMI) is a phenomenon observed from invertebrates to human. Memory extinction is proposed to be an active inhibitory modification of memory, however, whether extinction is affected in aging animals remains to be elucidated. Employing a modified paradigm for studying memory extinction in fruit flies, we found that only the stable, but not the labile memory component was suppressed by extinction, thus effectively resulting in higher memory loss in aging flies. Strikingly, young flies were able to fully restore the stable memory component 3 h post extinction, while aging flies failed to do so. In conclusion, our findings reveal that both accelerated extinction and impaired restoration contribute to memory impairment in aging animals.
Sigma-Aldrich
3-Octanol, natural, ≥97%, FCC, FG
Sigma-Aldrich
3-Octanol, ≥97%, FCC, FG
Sigma-Aldrich
4-Methylcyclohexanol, mixture of cis and trans, 98%
Sigma-Aldrich
3-Octanol, 99%