Merck
  • Toxicological screening of human plasma by on-line SPE-HPLC-DAD: identification and quantification of basic drugs and metabolites.
Toxicological screening of human plasma by on-line SPE-HPLC-DAD: identification and quantification of basic drugs and metabolites.
Biomedical chromatography : BMC (2014-11-18)
Ludmila Mut, Thomas Grobosch, Torsten Binscheck-Domaß, Wolfgang Frenzel
ABSTRACT
An automated multi-analyte screening method for the identification and quantification of 92 drugs and metabolites based on on-line solid-phase extraction-high-performance liquid chromatography-diode array detection technique was developed and successfully validated. In addition, a database with 870 entries including UV-spectra, relative/retention times and response factors of toxicologically relevant compounds was created. Plasma samples (0.2 mL) were treated with methanol, diluted with buffer and on-line extracted (Strata X, 20 ×2 mm, 25 µm) at pH 9. Analytical separation was carried out on a Gemini NX column (150 ×4.6 mm, 3 µm) using gradient elution with acetonitrile-water (90:10,v/v) and 0.05 m potassium dihydrogen phosphate buffer (pH 2.3). Linear calibration curves with correlation coefficients ≥0.9950 were obtained for 78 analytes. As an additional benefit, the newly developed method allows the quantification of 42 analytes (e.g. antidepressants, neuroleptics and anticonvulsants) in a concentration range suitable for therapeutic drug monitoring. Limits of quantitation ranged from 0.02 mg/L (chlordiazepoxide) to 3.4 mg/L (mexiletine). Inter- and intra-day precisions of quality control samples (low/high) were better than 15% (zolpidem) and accuracy (bias) ranged from -11% (opipramol, venlafaxine) to 11% (venlafaxine, trazodone). Tests for carry-over and sample stability under different storage conditions were also performed and stability was adequate. Four cases of poisoning analysis are presented.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Rufinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Mirtazapine, ≥98% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Ethyl-2-phenylmalonamide monohydrate, 99%
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
N-Desmethylclozapine
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
DPH, ≥98% (HPLC)