Merck
  • Interaction of intraocular lenses with fibronectin and human lens epithelial cells: Effect of chemical composition and aging.
Interaction of intraocular lenses with fibronectin and human lens epithelial cells: Effect of chemical composition and aging.
Journal of biomedical materials research. Part A (2015-06-27)
Lionel Tortolano, Carole Serrano, Emile Jubeli, Johanna Saunier, Najet Yagoubi
ABSTRACT
The aim of this study is to investigate in vitro interactions between hydrophobic acrylate intraocular lenses (IOLs) and their biological environment. The influence of lens chemical composition and aging on fibronectin (FN) adsorption and on IOLs cytotoxicity on human lens epithelial cells was examined. Cytotoxicity of acrylate monomers used in IOLs manufacture was also investigated. Four different IOLs were included in the study: Acrysof(®), Tecnis(®), EnVista(®), and iSert(®). Implants were artificially aged in a xenon arc chamber to simulate 2 years of light exposure. Fibronectin adsorption on IOL surface was quantified using ELISA and correlated to surface roughness determined with AFM. Direct contact cytotoxicity was determined with the MTT assay and cell morphology was observed with light microscopy. Results showed that fibronectin adsorption did not differ significantly among IOLs, whatever their chemical composition. Moreover, aging conditions did not impact fibronectin adsorption. All IOLs were biocompatible even after applying 2-year aging conditions, with cell viability higher than 70%. Five acrylate monomers appeared to be toxic in the range of concentrations tested, but no monomer release from the IOLs could be detected during accelerated 2-year incubation with saline solution. This study did not reveal an influence of chemical composition and aging on protein adsorption and on biocompatibility.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
2-Ethyl-2-phenylmalonamide monohydrate, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
1,4-Butanediol diacrylate, technical grade, contains ~75 ppm hydroquinone as inhibitor