Merck
  • Intratracheal administration of mitochondrial DNA directly provokes lung inflammation through the TLR9-p38 MAPK pathway.
Intratracheal administration of mitochondrial DNA directly provokes lung inflammation through the TLR9-p38 MAPK pathway.
Free radical biology & medicine (2015-03-17)
Xiaoling Gu, Guannan Wu, Yanwen Yao, Junli Zeng, Donghong Shi, Tangfeng Lv, Liang Luo, Yong Song
ABSTRACT
An increasing number of studies have focused on the phenomenon that mitochondrial DNA (mtDNA) activates innate immunity responses. However, the specific role of mtDNA in inflammatory lung disease remains elusive. This study was designed to examine the proinflammatory effects of mtDNA in lungs and to investigate the putative mechanisms. C57BL/6 mice were challenged intratracheally with mtDNA with or without pretreatment with chloroquine. Changes in pulmonary histopathology, cytokine concentrations, and phosphorylation levels of p38 MAPK were assayed at four time points. In in vitro experiments, THP-1 macrophages were pretreated or not pretreated with chloroquine, TLR9 siRNA, p38 MAPK siRNA, or SB203580 and then incubated with mtDNA. The levels of cytokines and p-p38 MAPK were detected by ELISA and Western blot, respectively. The intratracheal administration of mtDNA induced infiltration of inflammatory cells, production of proinflammatory cytokines (including IL-1β, IL-6, and TNF-α), and activation of p38 MAPK. The chloroquine pretreatment resulted in an abatement of mtDNA-induced local lung inflammation. In vitro experiments showed that the exposure of THP-1 macrophages to mtDNA also led to a significant upregulation of IL-1β, IL-6, and TNF-α and the activation of p38 MAPK. And these responses were inhibited either by chloroquine and TLR9 siRNA or by SB203580 and p38 MAPK siRNA pretreatment. The intratracheal administration of mtDNA induced a local inflammatory response in the mouse lung that depended on the interactions of mtDNA with TLR9 and may be correlated with infiltrating macrophages that could be activated by mtDNA exposure via the TLR9-p38 MAPK signal transduction pathway.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mapk14
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK14
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR9, RP11-330H6.5
Sigma-Aldrich
MISSION® esiRNA, targeting human TWF2, RP11-330H6.5