Merck
  • Enhanced red emission on co-doping of divalent ions (M(2+)=Ca(2+), Sr(2+), Ba(2+)) in YVO4:Eu(3+) phosphor and spectroscopic analysis for its application in display devices.
Enhanced red emission on co-doping of divalent ions (M(2+)=Ca(2+), Sr(2+), Ba(2+)) in YVO4:Eu(3+) phosphor and spectroscopic analysis for its application in display devices.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-07-22)
Puja Kumari, J Manam
ABSTRACT
This paper reports the enhancement in photoluminescence due to the alkaline ions (M(2+)) co-doping in YVO4:Eu(3+) phosphors prepared by solid state reaction method. The as synthesized samples were found to have single phase tetragonal structure and the incorporation of dopant/codopant did not affect the crystal structure. The characteristic bands of YO and VO were located at 450cm(-1) and 830cm(-1), respectively. The microscopic images exhibited the irregular morphology of the synthesized phosphor consisting of μm sized particles. Upon UV excitations, all phosphors showed intense red-orange emissions corresponding to the (5)D0→(7)FJ=1,2:Eu(3+) transitions, and the emission intensity was further enhanced due to codoping. Divalent ions introduced radiative defect centers, which enhances the PL intensity significantly. The calculation of spectral parameters was done through Judd-Ofelt theory from emission spectra. The photometric characterizations and corresponding calculations indicated the suitability of the phosphor for display devices.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%