Merck
  • An epimer of deoxynivalenol: purification and structure identification of 3-epi-deoxynivalenol.
An epimer of deoxynivalenol: purification and structure identification of 3-epi-deoxynivalenol.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-08-08)
Jian Wei He, Raymond Yang, Ting Zhou, Greg J Boland, Peter M Scott, Genevieve S Bondy
ABSTRACT
In an investigation of deoxynivalenol (DON)-transformation products by Devosia mutans 17-2-E-8, the major product was identified as 3-epi-DON. This DON-transformation product was analysed by liquid chromatography and identified by congruent retention time and UV/Vis spectrum, as well as mass spectrometric data. Nuclear magnetic resonance (NMR) experiments including correlation spectroscopy (COSY), heteronuclear single quantum coherence (HSQC) and nuclear overhauser effect (NOE) were conducted for structural characterisation of 3-epi-DON. High-speed counter-current chromatography (HSCCC) was applied to scale up the separation of 3-epi-DON from DON in a D. mutans 17-2-E-8 culture. From the culture where 100 mg DON was applied, 56 mg of 3-epi-DON (purity of 96.8%) was obtained from the HSCCC. The purified 3-epi-DON will be used for toxicological characterisation studies of this chemical.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl ether, ≥99%
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Dimethyl ether, ≥99.9%