Merck
  • Common-ion effects on the deliquescence lowering of crystalline ingredient blends.
Common-ion effects on the deliquescence lowering of crystalline ingredient blends.
Food chemistry (2015-11-18)
Matthew Allan, Lynne S Taylor, Lisa J Mauer
ABSTRACT
Deliquescence points (RH0, RH0mix) of ionic crystalline food ingredients and blends thereof were determined using water activity and moisture sorption techniques. Measured RH0mix values of ingredient blends with and without a common ion were compared to Ross equation predictions of deliquescence lowering. In binary blends with no common ion, measured RH0mix values ranged from 5% RH lower to 6% RH higher than predicted; however, when a common ion was present, the measured RH0mix was consistently 6-8% RH higher than predicted. In tertiary blends with a common ion, RH0mix values were 15-18% RH higher than predicted. The higher RH0mix in blends with a common ion is caused by counterion dissociation competition from the common ion. At equilibrium, these solutions will have fewer solutes and a greater vapor pressure than assumed by the Ross equation. A modified Ross equation was developed to compensate for the common-ion effect and improve RH0mix predictions.
Product Description
Supelco
Sodium Saccharin, analytical standard
Sigma-Aldrich
Calcium L-ascorbate dihydrate, 99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%