Merck
  • Characterization and immunomodulatory activity of rice hull polysaccharides.
Characterization and immunomodulatory activity of rice hull polysaccharides.
Carbohydrate polymers (2015-04-04)
Li-Chan Yang, Chang-Chi Hsieh, Wen-Chuan Lin
ABSTRACT
Rice hulls (Oryza sativa) are high in carbohydrate content and have been utilized as dietary fiber. The immunomodulatory bioactivity of rice hull polysaccharides (RHPS) has rarely been reported. This study demonstrated the structural characteristics and immunomodulating of RHPS. The RHPS were fractioned using DEAE-650M column, producing one neutral and 3 acidic polysaccharide fractions. RHPS were examined using HPAEC-PAD, HP-SEC, NMR and GC-MS for structural characteristics. The results showed that RHPS consisted of arabinose, galactose, glucose, mannose, and xylose in ratios of 10:44.8:29.8:9.3:6.1 and comprised (1→3)-Gal as backbone, and its average molecular weight was 77kDa. The presence of type II arabinogalactan (AGII) was confirmed through LM2-ELISA and Yariv gel diffusion showed the RHPS had AGII features. This study examined the immunomodulatory effects of orally administering RHPS in vivo. The RHPS increased the cytotoxicity of splenic natural killer cells, macrophage phagocytosis, and cytokine inductions. This is the first study to demonstrate the structural characteristics of an active polysaccharide from rice hulls and its immunopharmacological effects in vivo.
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Barium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)