Merck
  • A novel method for the synthesis of symmetrical triacylglycerols by enzymatic transesterification.
A novel method for the synthesis of symmetrical triacylglycerols by enzymatic transesterification.
Bioresource technology (2015-08-22)
Wenjia Tang, Xiaosan Wang, Jianhua Huang, Qingzhe Jin, Xingguo Wang
ABSTRACT
A novel two-step enzymatic method is described in this study to synthesize symmetrical triacylglycerols (TAGs) with arachidonic acid (ARA) at the sn-2 position. The processes included the synthesis of 2-monoacylglycerols (2-MAGs) rich in 2-arachidonoylglycerol (2-AG) by enzymatic ethanolysis and the synthesis of symmetrical TAGs by enzymatic transesterification between 2-MAGs and vinyl palmitate. Under the optimal conditions, desired symmetrical TAGs were obtained at 89% yield. In this study, vinyl palmitate rather than palmitic acid was used as a novel acyl donor to react with 2-MAGs. It was the first study reporting the synthesis of symmetrical TAGs by enzymatic transesterification. The reaction using fatty acid vinyl ester as acyl donor is irreversible and temperature is low. Low-temperature reaction greatly suppressed the acyl migration of 2-MAGs and the irreversible reaction is much more effective compared to reversible reactions using free fatty acid and fatty acid ester as acyl donors.
Sigma-Aldrich
1,3-Diolein, ≥99% (GC)
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Methyl arachidonate, ≥99% (GC)