Merck
Intestinal behavior of the ester prodrug tenofovir DF in humans.
International journal of pharmaceutics (2015-03-10)
Sophie Geboers, Steven Haenen, Raf Mols, Joachim Brouwers, Jan Tack, Pieter Annaert, Patrick Augustijns
ABSTRACT
Tenofovir-disoproxil-fumarate (TDF) is a double ester prodrug which enables intestinal uptake of tenofovir (TFV) after oral administration in humans. In this study, prodrug stability was monitored in situ in the human intestine and in vitro using biorelevant media. In fasted state human intestinal fluids, the prodrug was completely degraded within 90 min, resulting in the formation of the mono-ester intermediate and TFV; in fed state intestinal fluids, the degradation rate of TDF was slightly reduced and no TFV was formed. Intestinal fluid samples aspirated after administration of TDF confirmed extensive intraluminal degradation of TDF in fasted state conditions; a relatively fast absorption of TDF partly compensated for the degradation. Although food intake reduced intestinal degradation, the systemic exposure was not proportionally increased. The lower degradation in fed state conditions may be attributed to competing esterase substrates present in food, lower chemical degradation in the slightly more acidic environment and micellar entrapment, delaying exposure to the "degrading" intestinal environment. The results of this study demonstrate premature intestinal degradation of TDF and suggest that TFV may benefit from a more stable prodrug approach; however, fast absorption may compensate for fast degradation, indicating that prodrug selection should not be limited to stability assays.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Tetrabutylammonium sulfate solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%