Merck
  • Functional variations in MBL2 gene are associated with cutaneous leishmaniasis in the Amazonas state of Brazil.
Functional variations in MBL2 gene are associated with cutaneous leishmaniasis in the Amazonas state of Brazil.
Genes and immunity (2015-03-13)
F J de Araujo, T G Mesquita, L D O da Silva, S A de Almeida, W de S Vital, A Chrusciak-Talhari, J A de O Guerra, S Talhari, R Ramasawmy
ABSTRACT
Functional variations in the mannose-binding lectin (MBL2) gene causing low levels of serum MBL are associated with susceptibility to numerous infectious diseases. We investigated whether there is genetic association of MBL2 variant alleles with cutaneous leishmaniasis (CL) caused by Leishmania guyanensis. We used PCR-restriction fragment length polymorphism to genotype six MBL2 variants, three in the promoter region and three in the exon 1. An association was noted between the single nucleotide polymorphism -221X/Y of the MBL2 gene and CL (P=2.9 × 10(-6); odds ratio (OR)=1.9 (1.4-2.5) consistent with the hypothesis that the -221X allele confers high risk to development of CL among L. guyanensis-infected individuals. Furthermore, L. guyanensis-infected individuals bearing the codon 57 allele C had a higher risk of developing CL (P=5 × 10(-5); OR=1.9 (1.4-2.6)). The low MBL expressor haplotype LXPB was also associated to CL (P=6 × 10(-4)). This study raises the possibility that functional polymorphisms in MBL2 gene play a role in clinical outcome of Leishmania infection.
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Tin, foil, 1m coil, thickness 0.084mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 4mm disks, thickness 0.15mm, as rolled, 99.99+%
Tin, foil, 50mm disks, thickness 0.025mm, as rolled, 99.99+%
Tin, foil, 2m coil, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 25mm disks, thickness 1.5mm, as rolled, 99.99+%
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 99.99+%
Tin, foil, 4mm disks, thickness 0.004mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.030mm, as rolled, 97.4%
Tin, foil, 10mm disks, thickness 0.06mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 10mm disks, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 15mm disks, thickness 0.015mm, 99.75%
Tin, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.05mm, as rolled, 98.8%
Tin, foil, 15mm disks, thickness 0.004mm, 99.75%
Tin, foil, 15mm disks, thickness 0.06mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.75mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 0.075mm, as rolled, 98.8%