Merck
  • Optimization of low-temperature blanching combined with calcium treatment to inactivate Escherichia coli O157:H7 on fresh-cut spinach.
Optimization of low-temperature blanching combined with calcium treatment to inactivate Escherichia coli O157:H7 on fresh-cut spinach.
Journal of applied microbiology (2015-03-31)
N H Kim, N Y Lee, S H Kim, H J Lee, Y Kim, J H Ryu, M S Rhee
ABSTRACT
To develop a mild blanching method with calcium salts to ensure microbiological safety and quality of fresh-cut spinach. The antimicrobial efficacy of eight calcium salts was evaluated on Escherichia coli O157:H7 at 45-65°C and calcium hydroxide (Ca(OH)2 ) showed the greatest synergistic antimicrobial effect with heat. Combinations of low temperature treatments (45, 55, 65°C), time (20, 40, 60 s) and Ca(OH)2 (0·25, 0·50, 0·75%) were applied for E. coli O157:H7 disinfection on fresh-cut spinach to develop a predictive model using a Box-Behnken experimental design. A suitable quadratic model was produced (R(2) = 0·98, P < 0·001) and the optimum condition (64·9°C with 0·52% Ca(OH)2 for 42·4 s) was drawn by reducing 6·6 log CFU g(-1) of E. coli O157:H7 on fresh-cut spinach. Blanching at 61·9°C with 0·52% Ca(OH)2 for 41·7 s can inactivate E. coli O157:H7 on spinach by 5·4 log CFU g(-1) . The new method was comparable to the CDC recommendation for safe spinach cooking (71·1°C, 15 s; 5·0 log CFU g(-1) reduction) with lower levels of weight loss of the spinach (P < 0·05). This study suggests an efficient spinach blanching method for E. coli O157:H7 disinfection. This blanching method will enhance microbiological safety of fresh-cut produce while minimizing the use of energy and chemicals.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium propionate, 99.0-100.5% (in dried substance), meets analytical specification of E 282
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium propionate, ≥97.0% (dry substance, NT)
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)