Merck
  • UHPLC-PDA-ESI-TOF/MS metabolic profiling of Arctostaphylos pungens and Arctostaphylos uva-ursi. A comparative study of phenolic compounds from leaf methanolic extracts.
UHPLC-PDA-ESI-TOF/MS metabolic profiling of Arctostaphylos pungens and Arctostaphylos uva-ursi. A comparative study of phenolic compounds from leaf methanolic extracts.
Phytochemistry (2015-02-24)
Alessia Panusa, Rita Petrucci, Giancarlo Marrosu, Giuseppina Multari, Francesca Romana Gallo
ABSTRACT
The aim of this study was to get a rapid metabolic fingerprinting and to gain insight into the metabolic profiling of Arctostaphylos pungens H. B. K., a plant morphologically similar to Arctostaphylos uva-ursi (L.) Spreng. (bearberry) but with a lower arbutin (Arb) content. According to the European Pharmacopoeia the Arb content in the dried leaf of A. uva-ursi (L.) Spreng. must be at least 7% (wt/wt) but other species, like A. pungens, are unintentionally or fraudulently marketed instead of it. Therefore, methanolic leaf extracts of nine A. uva-ursi and six A. pungens samples labeled and marketed as "bearberry leaf" have been analyzed. A five-minute gradient with a UHPLC-PDA-ESI-TOF/MS on an Acquity BEH C18 (50×2.1 mm i.d.) 1.7 μm analytical column has been used for the purpose. A comprehensive assignment of secondary metabolites has been carried out in a comparative study of the two species. Among twenty-nine standards of natural compounds analyzed, fourteen have been identified, while other fifty-five metabolites have been tentatively assigned. Moreover, differences in both metabolic fingerprinting and profiling have been evidenced by statistical multivariate analysis. Specifically, main variations have been observed in the relative content for Arb, as expected, and for some galloyl derivative like tetra- and pentagalloylglucose more abundant in A. uva-ursi than in A. pungens. Furthermore, differences in flavonols profile, especially in myricetin and quercetin glycosilated derivatives, were observed. Based on principal component analysis myricetrin, together with a galloyl arbutin isomer and a disaccharide are herein proposed as distinctive metabolites for A. pungens.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
1,3,6-Trigalloylglucose, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Shikimic acid, ≥99%
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Ultrapure Acetonitrile