Merck
MiR-378b Promotes Differentiation of Keratinocytes through NKX3.1.
PloS one (2015-08-28)
Xi-Liang Wang, Tao Zhang, Jing Wang, Dian-Bao Zhang, Feng Zhao, Xue-Wen Lin, Zhe Wang, Ping Shi, Xi-Ning Pang
ABSTRACT
MicroRNA (miRNA) is a kind of short non-coding RNA, involved in various cellular processes. During keratinocyte differentiation, miRNAs act as important regulators. In this study, we demonstrated by microarray assay that the expression of miR-378b significantly increased during keratinocytes differentiation. Our findings showed that miR-378b could inhibit proliferation, migration and differentiation in keratinocytes. Luciferase reporter assays showed that miR-378b directly target NKX3.1. Silencing of NKX3.1 could coincide with the effects of miR-24 overexpression. In conclusion, our results demonstrate miR-378b promote keratinocytes differentiation by targeting NKX3.1. Manipulation of miR-378b may afford a new strategy to clinic treatment of skin injury and repair.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nkx3-1
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human NKX3-1
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis