Merck
  • Molecular characterization and expression analysis of mouse epidermal growth factor-like domain 8.
Molecular characterization and expression analysis of mouse epidermal growth factor-like domain 8.
International journal of molecular medicine (2015-06-23)
Ik-Jin Song, Muhammad Ikram, Fazli Subhan, Da-Jeong Choi, Ja-Rang Lee, Heui-Soo Kim, Young-Tak Lim, Sik Yoon
ABSTRACT
Epidermal growth factor (EGF)-like (EGFL) domain, a common structural module in numerous secreted or transmembrane proteins, is generally involved in protein-protein interactions. To date, several EGFL proteins have been identified and characterized, but little is known about EGFL domain 8 (EGFL8). The present study reported the molecular characterization and expression analysis of EGFL8 in mice. Mouse EGFL8 amplified using a reverse transcription-polymerase chain reaction approach was sequenced and characterized. Mouse EGFL8 encodes a protein of 293 amino acids with two EGFL domains, an Emilin-like domain and a Ca(2+)-binding EGFL domain, which has a molecular mass of 32 kDa. The coding sequence has a high degree of amino acid sequence identity across species, and the EGFL domain has been highly conserved in various species during evolutionary radiation. A phylogenetic tree calculated using the neighbor-joining method revealed that EGFL8 and EGFL7 are more closely associated with each other than either is to EGFL3, and they cluster with EGFL6. It was found that mouse EGFL8 protein was highly expressed in diverse mouse tissue types, including the thymus, lymph nodes, testis, ovaries, epididymis, ductus deferens, ileum, colon, stomach, esophagus, lung, uterus, urinary bladder, skin, spleen, adrenal glands and penis. These results are of great use in understanding the biological roles of mouse EGFL8 for further study.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Bicinchoninic Acid solution
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture