Merck
Chronic exposures to monomethyl phthalate in Western clawed frogs.
General and comparative endocrinology (2015-02-11)
Justine Mathieu-Denoncourt, Shane R de Solla, Valerie S Langlois
ABSTRACT
Polymer flexibility and elasticity is enhanced by plasticizers. However, plasticizers are often not covalently bound to plastics and thus can leach from products into the environment. Much research effort has focused on their effects in mammalian species, but data on aquatic species are scarce. In this study, Western clawed frog (Silurana tropicalis) embryos were exposed to 1.3, 12.3, and 128.7mg/L monomethyl phthalate (MMP) until the juvenile stage (11weeks) and to 1.3mg/L MMP until the adult stage (51weeks). MMP decreased survival, hastened metamorphosis, and biased the sex ratio toward males (2M:1F) at the juvenile stage without altering the expression of a subset of thyroid hormone-, sex steroid-, cellular stress- or transcription regulation-related genes in the juvenile frog livers. At the adult stage, exposure to MMP did not have significant adverse health effects, except that females had larger interocular distance and the expression of the heat shock protein 70 was decreased by 60% in the adult liver. In conclusion, this study shows that MMP is unlikely to threaten amphibian populations as only concentrations four orders of magnitude higher than the reported environmental concentrations altered the animal physiology. This is the first complete investigation of the effects of phthalates in a frog species, encompassing the entire life cycle of the organisms.
Sigma-Aldrich
Gentamicin sulfate from Micromonospora purpurea, ≥590 μg/mg of gentamicin
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~5,000 IU
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~10,000 IU
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, suitable for mouse embryo cell culture
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, from human pregnancy urine
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Calcium chloride