Merck
  • Gut-Associated Bacteria of Dendroctonus valens and their Involvement in Verbenone Production.
Gut-Associated Bacteria of Dendroctonus valens and their Involvement in Verbenone Production.
Microbial ecology (2015-05-20)
Letian Xu, Qiaozhe Lou, Chihang Cheng, Min Lu, Jianghua Sun
ABSTRACT
Bark beetles are the most important mortality agent in coniferous forests, and pheromones play important roles in their management. Dendroctonus valens LeConte was introduced from North America to China and has killed millions of healthy pines there. Trapping with semiochemicals and pheromones was deployed in D. valens management in the last decade, but little is known about the ability of gut bacteria to produce the pheromone. In this study, we analyzed the volatiles in D. valens guts and frass after antibiotic treatment versus control. Then, we isolated and identified the bacteria in D. valens guts and frass, examined verbenone (a multifunctional pheromone of D. valens) production by 16 gut bacterial isolates from the precursor cis-verbenol at three concentrations, and further compared the cytotoxicities between the cis-verbenol and verbenone to the bacterial isolates. cis-Verbenol was not detected in the frass in the control group, but it was in the antibiotic treatment. The amount of verbenone was significantly suppressed in D. valens guts after antibiotic treatment versus control. Thirteen out of 16 gut bacterial isolates were capable of cis-verbenol to verbenone conversion, and cis-verbenol had stronger cytotoxicities than verbenone to all tested gut bacterial isolates. The bacterial species capable of verbenone production largely exists in D. valens guts and frass, suggesting that gut-associated bacteria may help the bark beetle produce the pheromone verbenone in guts and frass. The bacteria may benefit from the conversion due to the reduced cytotoxicity from the precursor to the beetle pheromone.
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
(1S)-(−)-Verbenone, ≥93%
Sigma-Aldrich
(S)-cis-Verbenol, 95%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
(1S)-(−)-Verbenone, 94%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Heptyl acetate, ≥98%, FG
Sigma-Aldrich
Heptyl acetate, natural, ≥98%, FG