Merck
  • Detection of dihydroxycholesterols in human plasma using HPLC-ESI-MS/MS.
Detection of dihydroxycholesterols in human plasma using HPLC-ESI-MS/MS.
Steroids (2015-02-17)
Ratna Karuna, Isabelle Christen, Andreas W Sailer, Francis Bitsch, Juan Zhang
ABSTRACT
We report a straightforward sample preparation procedure and a direct liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the analysis of 7alpha,25-dihydroxycholesterol (7α25-OHC) and 7alpha,27-dihydroxycholesterol (7α27-OHC). By applying a slow protein precipitation approach using cold ethanol, we were able to detect and quantify 7α25-OHC and 7α27-OHC in a fast and reliable manner. The average concentrations from 20 healthy individuals were determined to be 0.21±0.05nM for 7α25-OHC and 3.4±0.1nM for 7α27-OHC. In addition, we are the first to report the average degrees of esterification (n=8) to be 73.8% and 82% for 7α25-OHC and 7α27-OHC, respectively. Using the established method, we achieved the sensitivity sufficient for detecting low abundant dihydroxylated oxysterols in healthy individuals. This result should enable extension of these studies towards a comprehensive analysis of oxysterol levels under disease conditions.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
7α,25-Dihydroxycholesterol, ≥98% (HPLC)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Avanti
7β,25-dihydroxycholesterol, Avanti Research - A Croda Brand
Avanti
7α,27-dihydroxycholesterol-d6, Avanti Research - A Croda Brand 700136P, powder
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, NMR reference standard
Avanti
7α,25-dihydroxycholesterol-d6, Avanti Research - A Croda Brand
Avanti
7α,27-dihydroxycholesterol, Avanti Research - A Croda Brand